Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIKFYVE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103242
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103242 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103242 Supplier Invitrogen™ Supplier No. PA5103242
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PIP5K.

PIKFYVE belongs to a large family of lipid kinases that alter the phosphorylation status of intracellular phosphatidylinositol. The content of phosphatidylinositol 3,5-bisphosphateP2) in endosomal membranes changes dynamically with fission and fusion events that generate or absorb intracellular transport vesicles. PIKFYVE is the PtdIns(3,5)P2-producing component of a trimolecular complex that tightly regulates the level of PtdIns(3,5)P2. Other components of this complex are the PIKFYVE activator VAC14 and the PtdIns(3,5)P2 phosphatase FIG4. In addition to its phosphoinositide 5-kinase activity, PIKFYVE also has protein kinase activity. For a review, see Shisheva.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIKFYVE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PIKFYVE
Gene Accession No. Q9Y2I7, Q9Z1T6
Gene Alias 1-phosphatidylinositol 3-phosphate 5-kinase; 1-phosphatidylinositol-3-phosphate 5-kinase; 5230400C17Rik; CFD; epididymis luminal protein 37; FAB1; FYVE finger-containing phosphoinositide kinase; HEL37; Kiaa0981; p235; Phosphatidylinositol 3-phosphate 5-kinase; Phosphatidylinositol 3-phosphate 5-kinase type III; phosphatidylinositol-3-phosphate 5-kinase type III; phosphatidylinositol-3-phosphate/phosphatidylinositol 5-kinase, type III; phosphatidylinositol-4-phosphate 5-kinase, type III; phosphoinositide kinase, FYVE finger containing; phosphoinositide kinase, FYVE-type zinc finger containing; PIKfyve; Pip5k; PIP5K3; Pipk5k3; PipkIII; PIPkin-III; PtdIns(4)P-5-kinase; type III PIP kinase; ZFYVE29; zinc finger, FYVE domain containing 29
Gene Symbols PIKFYVE
Host Species Rabbit
Immunogen A synthesized peptide derived from human PIKFYVE(Accession Q9Y2I7), corresponding to amino acid residues T283-N333.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18711, 200576
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.