Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Piko PCR Plate, 24-well, clear

Catalog No. spl0240
Change view
Click to view available options
Quantity:
200 plates
50 plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPL0240 200 plates
SPL024050 50 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SPL0240 Supplier Thermo Scientific™ Supplier No. SPL0240
Only null left
Add to Cart
Add to Cart

Thermo Scientific™ Piko PCR Plates are modular, ultra-thin wall PCR plates for Piko* Thermal Cyclers.

Thermo Scientific™ Piko™ PCR Plates are modular, ultra-thin wall PCR plates for Piko Thermal Cyclers.

Piko PCR Plates are also flexible alternatives for conventional microplates. Four plates, each the size of a microscope slide, can be reversibly snapped into a rigid frame, producing the equivalent of a standard 96- or 384-well microplate. This format allows multiple sample sources to be combined into a single PCR run, or subjected to different post-PCR processing.
  • Wall thickness half that of conventional thin-walled tubes
  • Flexible format saves cost and reduces waste
  • Compatible with standard multi-channel pipettes and liquid handlers
  • Well spacing and footprint correspond to industry (ANSI) dimensions
  • Available in clear and white

Recommended for:

PCR, Fast PCR, High Performance PCR

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified RNase/DNase-free
Packaging 200 Plate
Product Line Piko
Color Clear
Product Type PCR Microplate
For Use With (Equipment) Piko Thermal Cycler, PikoReal Real-Time PCR System
Material Polypropylene
No. of Wells 24
Quantity 200 plates
Reaction Speed Low profile
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.