Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foamtec PilloWIPE™ Wipers

Catalog No. p-4258176 Shop All Foamtec International Products
Click to view available options
Dimensions (L x W):
10 x 10 in. (25.4 x 25.4 cm)
4 x 3 in. (10.16 x 7.62 cm)
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed to enhance the cleaning of difficult to clean surfaces such as tools with sharp edges or high temperatures

Constructed from polyester and micro-fiber based, cleanroom-grade cloths and reticulated, cleanroom-grade foams.

  • Foam sponge interior provides cushioning for scratch sensitive surfaces such as transfer rollers and Electro static chucks
  • High absorbency and high tear strength make it perfect for heavy duty wiping applications
  • Sponge-like qualities allow screens, stencils, blenders, mixers and mills to be wiped clean with far less cost, cross-contamination and generation of hazardous waste
  • Sponge-like interior enables aggressive wiping while reducing the risk of generating fibers resulting from abrasion
  • Soft, pliable construction allows for wiping cloth to conform to articulated surfaces for thorough cleaning
  • Perfect for cleaning hot surfaces in wafer processing equipment such as shower heads

Specifications

Product Type Critical Task Cleanroom Wipe
Length (English) 4 in.
Length (Metric) 10.16 cm
Width (English) 3 in.
Width (Metric) 7.62 cm
Material Polyester
Absorbancy Highly absorbent
Autoclavable Autoclavable
Construction MiraWIPE™ outer Fabric, Sandwiched Foam sponge with netting and soft and pliable construction
Dimensions (L x W) 4 x 3 in. (10.16 x 7.62 cm)
For Use With (Application) Designed to enhance the cleaning of difficult to clean surfaces such as tools with sharp edges or high temperatures
Sterility Sterile
Thickness (English) 0.375 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.