Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PILR-alpha Antibody (2175D), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB64841J
Description
PILR-alpha Monoclonal antibody specifically detects PILR-alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
PILR-alpha | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry | |
2175D | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha | |
Mouse myeloma cell line NS0-derived recombinant human PILR-alpha with a C-terminal 6-His tag, Gln20-Thr196, Accession # Q9UKJ1 | |
0.1 mL | |
Signal Transduction | |
29992 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction