Learn More
                                    PILR-alpha Antibody (2175D), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB64841MFV450
Description
PILR-alpha Monoclonal antibody specifically detects PILR-alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
| PILR-alpha | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Signal Transduction | |
| 29992 | |
| Store at 4C in the dark. | 
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 2175D | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha | |
| Mouse myeloma cell line NS0-derived recombinant human PILR-alpha with a C-terminal 6-His tag, Gln20-Thr196, Accession # Q9UKJ1 | |
| RUO | |
| Primary | |
| Human | |
| IgG | 