Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PILRA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566000
Description
Immunogen sequence: ALSSSTSPRAP PSHRPLKSPQ NETLYSVLK Highest antigen sequence identity to the following orthologs - mouse 50%, rat 57%.
SHP-1-mediated dephosphorylation of protein tyrosine residues is central to the regulation of several cell signaling pathways. Two types of inhibitory receptor superfamily members are immunoreceptor tyrosine-based inhibitory motif (ITIM)-bearing receptors and their non-ITIM-bearing, activating counterparts. Control of cell signaling via SHP-1 is thought to occur through a balance between PILRalpha-mediated inhibition and PILR beta-mediated activation. This particular gene encodes the ITIM-bearing member of the receptor pair, which functions in the inhibitory role.Cell signaling pathways rely on a dynamic interaction between activating and inhibiting processes. SHP-1-mediated dephosphorylation of protein tyrosine residues is central to the regulation of several cell signaling pathways. Two types of inhibitory receptor superfamily members are immunoreceptor tyrosine-based inhibitory motif (ITIM)-bearing receptors and their non-ITIM-bearing, activating counterparts. Control of cell signaling via SHP-1 is thought to occur through a balance between PILR alpha-mediated inhibition and PILRbeta-mediated activation. These paired immunoglobulin-like receptor genes are located in a tandem head-to-tail orientation on chromosome 7. This particular gene encodes the ITIM-bearing member of the receptor pair, which functions in the inhibitory role.
Specifications
PILRA | |
Polyclonal | |
Unconjugated | |
PILRA | |
AV021745; cell surface receptor FDF03; FDF03; Inhibitory receptor PILR-alpha; paired immunoglobin like type 2 receptor alpha; paired immunoglobin-like type 2 receptor alpha; paired immunoglobulin-like type 2 receptor alpha; Pilra | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29992 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKJ1 | |
PILRA | |
Recombinant Human PILRA. Recombinant protein control fragment (Product #RP-106688). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction