Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIM1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | PIM1 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 5 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP231366SS
![]() |
Novus Biologicals
NBP231366SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP231366
![]() |
Novus Biologicals
NBP231366 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
PIM1 Polyclonal specifically detects PIM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PIM1 | |
Western Blot 2 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
P11309, P26794 | |
5292 | |
Partial recombinant protein made to an N-terminal portion of human PIM1 (between residues 1-200) [UniProt# P11309] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
45.4 kDa |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Protein Kinase, Tumor Suppressors | |
PBS with 0.05% Sodium Azide | |
EC 2.7.11.1, Oncogene PIM1, PIM, pim-1 kinase 44 kDa isoform, pim-1 oncogene, pim-1 oncogene (proviral integration site 1), proto-oncogene serine/threonine-protein kinase pim-1 | |
PIM1 | |
IgG | |
Protein A purified | |
This antibody may detect isoform 1 as well as isoform 2 of PIM1 (Uniprot # P11309). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title