Learn More
Promega PIM1 Kinase Enzyme System, 10microg
Catalog No. PRV4032 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4032 10 μg
1 options
Catalog No. PRV4032 Supplier Promega Supplier No. V4032
Recombinant full-length human PIM1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PIM1 is a proto-oncogene that belongs to a family of serine/threonine protein kinases that are highly conserved in multicellular organisms.
  • Easily Screen and Profile PIM1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C