Learn More
Promega PIM1 Kinase Enzyme System, 1mg
Catalog No. PRV6471 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6471 1 mg
1 options
Catalog No. PRV6471 Supplier Promega Supplier No. V6471
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human PIM1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PIM1 is a proto-oncogene that belongs to a family of serine/threonine protein kinases that are highly conserved in multicellular organisms.
  • Easily Screen and Profile PIM1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C