Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIM1 Monoclonal Antibody (G.360.1)
Rabbit Monoclonal Antibody
Supplier: Invitrogen™ MA514802
Description
It is not recommended to aliquot this antibody.
The proto-oncogene PIM1 is upregulated in prostate cancer. PIM1, which belongs to the Serine/Threonine protein kinase family, is thought to play a role in signal transduction in blood cells. The protooncogene PIM1 encodes a protein kinase upregulated in prostate cancer. It may affect the structure or silencing of chromatin by phosphorylating HP1 gamma/CBX3. PIM1 is expressed primarily in cells of the hematopoietic and germ line lineages.
Specifications
PIM1 | |
Monoclonal | |
74 μg/mL | |
0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
P06803, P11309, P26794, Q9N0P9 | |
PIM1 | |
Synthetic peptide corresponding to residues surrounding Val160 of Pim-1. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey, Bovine | |
Antibody | |
IgG |
Western Blot | |
G.360.1 | |
Unconjugated | |
PIM1 | |
non-specific serine/threonine protein kinase; Oncogene PIM1; PIM; Pim1; Pim-1; pim-1 kinase 44 kDa isoform; pim-1 oncogene; pim-1 oncogene (proviral integration site 1); Pim-1 proto-oncogene, serine/threonine kinase; proto-oncogene serine/threonine-protein kinase Pim-1; proviral integration site 1; serine/threonine-protein kinase pim-1; serine-threonine kinase PIM-1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18712, 24649, 281402, 5292 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction