Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIM1 Monoclonal Antibody (ZP003)
GREENER_CHOICE

Catalog No. 394600
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-460-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-460-0 Supplier Invitrogen™ Supplier No. 394600
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

42-4800 has been successfully used in ChIP analysis of PIM1 in human J9.2 J-lat T lymphocytes.

The proto-oncogene PIM1 is upregulated in prostate cancer. PIM1, which belongs to the Serine/Threonine protein kinase family, is thought to play a role in signal transduction in blood cells. The protooncogene PIM1 encodes a protein kinase upregulated in prostate cancer. It may affect the structure or silencing of chromatin by phosphorylating HP1 gamma/CBX3. PIM1 is expressed primarily in cells of the hematopoietic and germ line lineages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIM1
Applications ChIP Assay, ELISA, Western Blot
Classification Monoclonal
Clone ZP003
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene PIM1
Gene Accession No. P11309
Gene Alias non-specific serine/threonine protein kinase; Oncogene PIM1; PIM; Pim1; Pim-1; pim-1 kinase 44 kDa isoform; pim-1 oncogene; pim-1 oncogene (proviral integration site 1); Pim-1 proto-oncogene, serine/threonine kinase; proto-oncogene serine/threonine-protein kinase Pim-1; proviral integration site 1; serine/threonine-protein kinase pim-1; serine-threonine kinase PIM-1
Gene Symbols PIM1
Host Species Mouse
Immunogen Synthetic peptide derived from the internal region of the human Pim-1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5292
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.