Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536392
Description
This antibody detects endogenous protein at a molecular weight of 45 kDa. Purity is >95% by SDS-PAGE.
The proto-oncogene PIM1 is upregulated in prostate cancer. PIM1, which belongs to the Serine/Threonine protein kinase family, is thought to play a role in signal transduction in blood cells. The protooncogene PIM1 encodes a protein kinase upregulated in prostate cancer. It may affect the structure or silencing of chromatin by phosphorylating HP1 gamma/CBX3. PIM1 is expressed primarily in cells of the hematopoietic and germ line lineages.
Specifications
PIM1 | |
Polyclonal | |
Unconjugated | |
PIM1 | |
non-specific serine/threonine protein kinase; Oncogene PIM1; PIM; Pim1; Pim-1; pim-1 kinase 44 kDa isoform; pim-1 oncogene; pim-1 oncogene (proviral integration site 1); Pim-1 proto-oncogene, serine/threonine kinase; proto-oncogene serine/threonine-protein kinase Pim-1; proviral integration site 1; serine/threonine-protein kinase pim-1; serine-threonine kinase PIM-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18712, 24649, 5292 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P06803, P11309, P26794 | |
PIM1 | |
Synthetic peptide corresponding to amino acids 271-320 of human Pim-1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction