Learn More
Promega PIM2 Kinase Enzyme System, 10microg
Catalog No. PRV4034 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4034 10 μg
1 options
Catalog No. PRV4034 Supplier Promega Supplier No. V4034
Recombinant full-length human PIM2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PIM2 is a serine threonine kinase that is present in all tissues.
  • Easily Screen and Profile PIM2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C