Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIM2 Monoclonal Antibody (CHU61), Active Motif™

Rat Monoclonal Antibody
$132.55 - $397.65
Specifications
Antigen | PIM2 |
---|---|
Clone | CHU61 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3197
![]() ![]() |
Active Motif
61160 |
10 μg |
Each for $132.55
|
|
|||||
50-199-3196
![]() ![]() |
Active Motif
61159 |
100 μg |
Each for $397.65
|
|
|||||
Description
PIM proteins (PIM-1, PIM-2 and PIM-3) are oncogene-encoded serine/threonine kinases. PIM-2 is highly homologous to Pim-1 with similar oncogenic functions. PIM-2 is most abundantly expressed in hematopoietic tissues, spleen, thymus, and peripheral blood leukocytes, as well as in testis, small intestine, and colon. PIM-2 overexpression promotes resistance to a host of apoptotic stimuli; its expression is negatively regulated by growth factor depletion. Increased levels of PIM-2 has also been observed in certain cancers.Specifications
PIM2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
DXCch3; PIM2; Pim-2; pim-2 oncogene; Pim-2 proto-oncogene, serine/threonine kinase; Pim-2h; proto-oncogene Pim-2 (serine threonine kinase); proviral integration site 2; serine/threonine protein kinase pim-2; Serine/threonine-protein kinase pim-2 | |
PIM2 | |
IgG2a | |
Protein G | |
Antibody |
CHU61 | |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rat | |
Human | |
Q9P1W9 | |
11040 | |
This PIM2 antibody was raised against a recombinant protein corresponding to human PIM2. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
PIM2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title