Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIM2 Monoclonal Antibody (OTI5B5), TrueMAB™, OriGene
SDP

Catalog No. 501672204
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-2204 100 μL
50-254-2566 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-2204 Supplier OriGene Supplier No. TA501061
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PIM proteins (PIM-1, PIM-2 and PIM-3) are oncogene-encoded serine/threonine kinases. PIM-2 is highly homologous to Pim-1 with similar oncogenic functions. PIM-2 is most abundantly expressed in hematopoietic tissues, spleen, thymus, and peripheral blood leukocytes, as well as in testis, small intestine, and colon. PIM-2 overexpression promotes resistance to a host of apoptotic stimuli; its expression is negatively regulated by growth factor depletion. Increased levels of PIM-2 has also been observed in certain cancers.

Specifications

Antigen PIM2
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone OTI5B5
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PIM2
Gene Accession No. Q9P1W9
Gene Alias Pim-2 proto-oncogene, serine/threonine kinase
Gene Symbols PIM2
Host Species Mouse
Immunogen Full length human recombit protein of human PIM2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11040
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.