Learn More
Thermo Scientific Chemicals Pimecrolimus
$369.00 - $1079.00
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AC464991000
|
Thermo Scientific Chemicals
464991000 |
100 mg |
Each for $369.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AC464995000
|
Thermo Scientific Chemicals
464995000 |
500 mg |
Each for $1,079.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
137071-32-0 | |
C43 H68 Cl N O11 |
Specifications
White to beige | |
Glass Bottle | |
137071-32-0 | |
normal conditions | |
Pimecrolimus | |
Powder |
≥96.0% by HPLC | |
C43 H68 Cl N O11 | |
Refrigerator +4°C | |
synthetic source | |
810.45 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.