Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pimobendan, EP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18605226 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-226 25 g Amber Glass Bottle
18-605-224 1 g Amber Glass Bottle
18-605-225 5 g Amber Glass Bottle
18-605-227 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-226 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P198225GM
Add to Cart
Add to Cart
Request a Quote

P1982, 74150-27-9, C19H18N4O2

Spectrum™ Chemical Pimobendan, EP is used for the management of congestive heart failure. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 74150-27-9
Molecular Formula C19H18N4O2
Molecular Weight (g/mol) 334.38
InChI Key GLBJJMFZWDBELO-UHFFFAOYNA-N
IUPAC Name 6-[2-(4-methoxyphenyl)-1H-1,3-benzodiazol-6-yl]-5-methyl-2,3,4,5-tetrahydropyridazin-3-one
SMILES COC1=CC=C(C=C1)C1=NC2=CC=C(C=C2N1)C1=NNC(=O)CC1C

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98 to 101%
Grade EP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.