Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pimobendan, Thermo Scientific™

Catalog No. AC468570010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468570010 1 g
AC468572500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468570010 Supplier Thermo Scientific Chemicals Supplier No. 468570010
Add to Cart

Chemical Identifiers

CAS 74150-27-9
Molecular Formula C19H18N4O2
Molecular Weight (g/mol) 334.38
MDL Number 00761648
InChI Key GLBJJMFZWDBELO-UHFFFAOYNA-N
IUPAC Name 6-[2-(4-methoxyphenyl)-1H-1,3-benzodiazol-6-yl]-5-methyl-2,3,4,5-tetrahydropyridazin-3-one
SMILES COC1=CC=C(C=C1)C1=NC2=CC=C(C=C2N1)C1=NNC(=O)CC1C

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Concentration or Composition Notes Titration with HClO4
Physical Form Powder
Concentration 97.5% to 102.5%
UN Number 2811
Color White to Light Yellow
Merck Index 7516
Quantity 1 g
Formula Weight 334.37
Chemical Name or Material Pimobendan
Show More Show Less
Hazard Category GHS07: Exclamation mark
ShelfLife 3 years
RTECSNumber UR6373000
Recommended Storage Refrigerator +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.