Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Pimonidazole
GREENER_CHOICESDP

Catalog No. 618250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-825-0 50 mg
61-821-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-825-0 Supplier Tocris Bioscience™ Supplier No. 6182/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Hypoxia detection reagent

Pimonidazole is a hypoxia detection reagent. Labels hypoxic tumor cells in vivo. Also labels hypoxic mesenchymal stem cells in mouse bone marrow.

Specifications

Chemical Name or Material α-[(2-Nitro-1H-imidazol-1-yl)methyl]-1-piperidineethanol
CAS 70132-50-2
Assay Percent Range >98%
Quantity 50 mg
Molecular Formula C11H18N4O3
Formula Weight Observed MW: 254.29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.