Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543252
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543252 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543252 Supplier Invitrogen™ Supplier No. PA543252
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IDENPASDFD DSGSLLGFKY GSGQKYGGIP NFSHRQVRYL EKNVKLKSKY Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 93%; Horse: 85%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.

Catalyzes the 2 serial methylation steps for the conversion of the 7-monomethylguanosine (m(7)G) caps of snRNAs and snoRNAs to a 2,2,7-trimethylguanosine (m(2,2,7)G) cap structure. The enzyme is specific for guanine, and N7 methylation must precede N2 methylation. Hypermethylation of the m7G cap of U snRNAs leads to their concentration in nuclear foci, their colocalization with coilin and the formation of canonical Cajal bodies (CBs). Plays a role in transcriptional regulation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIMT
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TGS1
Gene Accession No. Q96RS0
Gene Alias Cap-specific guanine-N2 methyltransferase; CLL-associated antigen KW-2; D4Ertd800e; HCA137; Hepatocellular carcinoma-associated antigen 137; LOW QUALITY PROTEIN: trimethylguanosine synthase; NCOA6IP; nuclear receptor coactivator 6 interacting protein; nuclear receptor coactivator 6-interacting protein; Pimt; PIPMT; PRIP-interacting protein with methyltransferase motif; TGS1; tgs1 {ECO:0000250; Trimethylguanosine synthase; trimethylguanosine synthase 1; trimethylguanosine synthase homolog; trimethylguanosine synthase homolog (S. cerevisiae); trimethylguanosine synthase-like protein; UniProtKB:Q96RS0}
Gene Symbols TGS1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TGS1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 96764
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.