Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576285
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Catalyzes the 2 serial methylation steps for the conversion of the 7-monomethylguanosine (m(7)G) caps of snRNAs and snoRNAs to a 2,2,7-trimethylguanosine (m(2,2,7)G) cap structure. The enzyme is specific for guanine, and N7 methylation must precede N2 methylation. Hypermethylation of the m7G cap of U snRNAs leads to their concentration in nuclear foci, their colocalization with coilin and the formation of canonical Cajal bodies (CBs). Plays a role in transcriptional regulation. [UniProt].
Specifications
PIMT | |
Polyclonal | |
Unconjugated | |
TGS1 | |
Cap-specific guanine-N2 methyltransferase; CLL-associated antigen KW-2; D4Ertd800e; HCA137; Hepatocellular carcinoma-associated antigen 137; LOW QUALITY PROTEIN: trimethylguanosine synthase; NCOA6IP; nuclear receptor coactivator 6 interacting protein; nuclear receptor coactivator 6-interacting protein; Pimt; PIPMT; PRIP-interacting protein with methyltransferase motif; TGS1; tgs1 {ECO:0000250; Trimethylguanosine synthase; trimethylguanosine synthase 1; trimethylguanosine synthase homolog; trimethylguanosine synthase homolog (S. cerevisiae); trimethylguanosine synthase-like protein; UniProtKB:Q96RS0} | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116940, 96764 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q923W1, Q96RS0 | |
TGS1 | |
Recombinant fusion protein of human TGS1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction