Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIN/DLC8 Rabbit anti-Human, Mouse, Rat, Clone: 3Q1J6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PIN/DLC8 |
---|---|
Clone | 3Q1J6 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
PIN/DLC8 Monoclonal antibody specifically detects PIN/DLC8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PIN/DLC8 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
cytoplasmic dynein light polypeptide, DLC1DNCLC1, DLC8MGC126137, DNCL1MGC126138, dynein light chain 1, cytoplasmic, Dynein light chain LC8-type 1, dynein, cytoplasmic, light polypeptide 1, dynein, light chain, LC8-type 1,8 kDa dynein light chain, hdlc1, LC8, PINLC8a, Protein inhibitor of neuronal nitric oxide synthase | |
A synthetic peptide corresponding to a sequence within amino acids 1-89 of human PIN/DLC8 (P63167). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
3Q1J6 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
p53 Pathway | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
8655 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title