Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pin1 Antibody (3G8), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP122956MFV450
Description
Pin1 Monoclonal antibody specifically detects Pin1 in Human samples. It is validated for Flow CytometrySpecifications
Pin1 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
3G8 | |
Flow Cytometry | |
dod, EC 5.2.1.8, peptidyl-prolyl cis/trans isomerase, NIMA-interacting, peptidylprolyl cis/trans isomerase, NIMA-interacting 1, peptidyl-prolyl cis-trans isomerase NIMA-interacting 1, Peptidyl-prolyl cis-trans isomerase Pin1, PPIase Pin1, prolyl isomerase, protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1, Rotamase Pin1, UBL5 | |
Recombinant human Pin1 (1-163 aa) purified from E. coli | |
0.1 mL | |
Cell Cycle and Replication, Mitotic Regulators, Phospho Specific | |
5300 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction