Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pin1 Antibody (3G8), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP122956PECY55
Description
Pin1 Monoclonal specifically detects Pin1 in Human samples. It is validated for Flow Cytometry.Specifications
Pin1 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
PIN1 | |
Recombinant human Pin1 (1-163 aa) purified from E. coli | |
0.1 mL | |
Primary | |
Anti-human Pin1 mAb, clone 3G8, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Pin1 protein. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
3G8 | |
Flow Cytometry | |
dod, EC 5.2.1.8, peptidyl-prolyl cis/trans isomerase, NIMA-interacting, peptidylprolyl cis/trans isomerase, NIMA-interacting 1, peptidyl-prolyl cis-trans isomerase NIMA-interacting 1, Peptidyl-prolyl cis-trans isomerase Pin1, PPIase Pin1, prolyl isomerase, protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1, Rotamase Pin1, UBL5 | |
Mouse | |
Protein G purified | |
Cell Cycle and Replication, Mitotic Regulators, Phospho Specific | |
5300.0 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction