Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pin1 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128699IR
Description
Pin1 Polyclonal antibody specifically detects Pin1 in Human samples. It is validated for Western BlotSpecifications
Pin1 | |
Polyclonal | |
Western Blot | |
dod, EC 5.2.1.8, peptidyl-prolyl cis/trans isomerase, NIMA-interacting, peptidylprolyl cis/trans isomerase, NIMA-interacting 1, peptidyl-prolyl cis-trans isomerase NIMA-interacting 1, Peptidyl-prolyl cis-trans isomerase Pin1, PPIase Pin1, prolyl isomerase, protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1, Rotamase Pin1, UBL5 | |
The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1 using the numbering given in entry NP_006212.1 (GeneID 5300). | |
0.1 mL | |
Cell Cycle and Replication, Mitotic Regulators, Phospho Specific | |
5300 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction