Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pin1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177757
Description
Pin1 Polyclonal specifically detects Pin1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation.Specifications
Pin1 | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
dod, EC 5.2.1.8, peptidyl-prolyl cis/trans isomerase, NIMA-interacting, peptidylprolyl cis/trans isomerase, NIMA-interacting 1, peptidyl-prolyl cis-trans isomerase NIMA-interacting 1, Peptidyl-prolyl cis-trans isomerase Pin1, PPIase Pin1, prolyl isomerase, protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1, Rotamase Pin1, UBL5 | |
Rabbit | |
18.24 kDa | |
0.1 mg | |
Cell Cycle and Replication, Mitotic Regulators | |
5300 | |
Human, Bovine, Canine, Monkey | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
1.1 mg/ml | |
Western Blot 1:500-1:3000, ELISA 1:2500-1:10000, Immunohistochemistry 5 ug/ml, Immunoprecipitation 1:100 | |
Q13526, Q13526, Q13526, Q13526 | |
PIN1 | |
This affinity purified Pin1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal sequence of human Pin1. (Uniprot: Q13526) | |
Affinity Purified | |
RUO | |
Primary | |
A BLAST analysis was used to suggest cross-reactivity with Pin1 from human, dog, bovine and monkey based on a 100% homology with the immunizing sequence. Expect partial reactivity with Pin1 from mouse and rat sources based on 92% sequence homologies. Reactivity against homologues from other sources is not known. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction