Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518594
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518594 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518594 Supplier Invitrogen™ Supplier No. PA518594
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, human and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

PIN1 (Peptidylprolyl Cis/Trans Isomerase, NIMA-Interacting 1) is a Protein Coding gene. Diseases associated with PIN1 include Meibomian Cyst and Viral Exanthem. Among its related pathways are Transport of the SLBP independent Mature mRNA and Cytokine Signaling in Immune system. Gene Ontology (GO) annotations related to this gene include peptidyl-prolyl cis-trans isomerase activity and mitogen-activated protein kinase binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIN1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Pin1
Gene Accession No. Q13526
Gene Alias 0610025L01Rik; D9Bwg1161e; DOD; EC 5.2.1.8; NIMA-interacting; peptidylprolyl cis/trans isomerase, NIMA-interacting 1; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 a; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 S homeolog; Peptidyl-prolyl cis-trans isomerase NIMA-interacting 1; peptidyl-prolyl cis-trans isomerase Pin1; pin1; pin1.S; pin1a; pin1-a; pin1-b; PPIase Pin1; prolyl isomerase; prolyl isomerase Pin1; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1; protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1; rotamase Pin1; ubl5; XELAEV_18021616mg
Gene Symbols Pin1
Host Species Goat
Immunogen Synthetic peptide sequence (KHSQSRRPSSWRQEKITR) corresponding to the internal amino acids of PIN1 (aa 63-80).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298696, 5300
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.