Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596057
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596057 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596057 Supplier Invitrogen™ Supplier No. PA596057
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MADEEKLPPG WEKRMSRSSG RVYYFNHITN ASQWERPSGN SSSGGKNGQG EPARVRCSHL LVKHSQSRRP SSWRQEKITR TKEEALELIN GYIQKIKSGE EDFESLASQF SDCSSAKARG DLGAFSRGQM QKPFEDASFA LRTGEMSGPV FTDSGIHIIL RTE; Positive Samples: PC-3, 293T, HeLa, Mouse kidney, Rat brain, Rat heart, Mouse brain; Cellular Location: Cytoplasm, Nucleus, Nucleus speckle.

PIN1 (Peptidylprolyl Cis/Trans Isomerase, NIMA-Interacting 1) is a Protein Coding gene. Diseases associated with PIN1 include Meibomian Cyst and Viral Exanthem. Among its related pathways are Transport of the SLBP independent Mature mRNA and Cytokine Signaling in Immune system. Gene Ontology (GO) annotations related to this gene include peptidyl-prolyl cis-trans isomerase activity and mitogen-activated protein kinase binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Pin1
Gene Accession No. Q13526, Q9QUR7
Gene Alias 0610025L01Rik; D9Bwg1161e; DOD; EC 5.2.1.8; NIMA-interacting; peptidylprolyl cis/trans isomerase, NIMA-interacting 1; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 a; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 S homeolog; Peptidyl-prolyl cis-trans isomerase NIMA-interacting 1; peptidyl-prolyl cis-trans isomerase Pin1; pin1; pin1.S; pin1a; pin1-a; pin1-b; PPIase Pin1; prolyl isomerase; prolyl isomerase Pin1; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1; protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1; rotamase Pin1; ubl5; XELAEV_18021616mg
Gene Symbols Pin1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-163 of human Pin1 (NP_0062121).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23988, 298696, 5300
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.