Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIN4 Antibody (14A7), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | PIN4 |
---|---|
Clone | 14A7 |
Dilution | Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
PIN4 Monoclonal antibody specifically detects PIN4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
PIN4 | |
Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
4 (parvulin), MGC138486, Parvulin-17, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin) | |
A synthesized peptide derived from Human PIN4 [UniProt Q9Y237] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
14A7 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
metabolism | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5303 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title