Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIN4 Antibody (OTI7H2), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273416B
Description
PIN4 Monoclonal specifically detects PIN4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PIN4 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
PIN4 | |
Full length recombinant protein of human PIN4 (NP_006214) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI7H2 | |
Western Blot, Immunohistochemistry | |
4 (parvulin), MGC138486, Parvulin-17, protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin) | |
Mouse | |
Affinity Purified | |
metabolism | |
5303.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction