Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598831
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598831 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598831 Supplier Invitrogen™ Supplier No. PA598831
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIN4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene encodes a member of the parvulin subfamily of the peptidyl-prolyl cis/trans isomerase protein family. The encoded protein catalyzes the isomerization of peptidylprolyl bonds, and may play a role in the cell cycle, chromatin remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIN4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Pin4
Gene Accession No. Q9CWW6, Q9Y237
Gene Alias 2410002I22Rik; EPVH; eukaryotic parvulin homolog; hEPVH; hPar14; hPar17; MGC138486; Par14; Par17; parvulin; parvulin-14; Parvulin-17; peptidyl-prolyl cis/trans isomerase EPVH; peptidylprolyl cis/trans isomerase NIMA-interacting, 4; peptidylprolyl cis/trans isomerase, NIMA-interacting 4; peptidyl-prolyl cis-trans isomerase NIMA-interacting 4; Peptidyl-prolyl cis-trans isomerase Pin4; PIN4; PPIase Pin4; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin); protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin); rotamase PIN4
Gene Symbols Pin4
Host Species Rabbit
Immunogen Recombinant Human Peptidyl-prolyl cis-trans isomerase NIMA-interacting 4 protein (1-131AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5303, 684441, 69713
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.