Learn More
PIN4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305660AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, X. tropicalis and Bovine
This gene encodes a member of the parvulin subfamily of the peptidyl-prolyl cis/trans isomerase protein family. The encoded protein catalyzes the isomerization of peptidylprolyl bonds, and may play a role in the cell cycle, chromatin remodeling.Specifications
PIN4 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9Y237 | |
PIN4 | |
Between 81 and 131 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PIN4 | |
Protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin), EPVH, MGC138486, PAR14, PAR17 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5303 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.