Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIN4 Recombinant Rabbit Monoclonal Antibody (14A7)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550059
Description
PIN4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
This gene encodes a member of the parvulin subfamily of the peptidyl-prolyl cis/trans isomerase protein family. The encoded protein catalyzes the isomerization of peptidylprolyl bonds, and may play a role in the cell cycle, chromatin remodeling.
Specifications
PIN4 | |
Recombinant Monoclonal | |
2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9Y237 | |
Pin4 | |
A synthesized peptide derived from human PIN4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
14A7 | |
Unconjugated | |
Pin4 | |
2410002I22Rik; EPVH; eukaryotic parvulin homolog; hEPVH; hPar14; hPar17; MGC138486; Par14; Par17; parvulin; parvulin-14; Parvulin-17; peptidyl-prolyl cis/trans isomerase EPVH; peptidylprolyl cis/trans isomerase NIMA-interacting, 4; peptidylprolyl cis/trans isomerase, NIMA-interacting 4; peptidyl-prolyl cis-trans isomerase NIMA-interacting 4; Peptidyl-prolyl cis-trans isomerase Pin4; PIN4; PPIase Pin4; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin); protein (peptidylprolyl cis/trans isomerase) NIMA-interacting, 4 (parvulin); rotamase PIN4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
5303 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction