Learn More
Pinacol, 99%
Description
Pinacol is used in the preparation of synthetic intermediates like pinacolborane, bis(pinacolato)diboron and pinacolchloroborane by reaction with boron trichloride. Further, it plays an important role as an intermediate for biologically active compounds like antiviral, antibacterial, antifungal and antituberculous.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pinacol is used in the preparation of synthetic intermediates like pinacolborane, bis(pinacolato)diboron and pinacolchloroborane by reaction with boron trichloride. Further, it plays an important role as an intermediate for biologically active compounds like antiviral, antibacterial, antifungal and antituberculous.
Solubility
Soluble in hot water, alcohol, and diethyl ether.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong bases and strong acids.
Specifications
Specifications
| Melting Point | 42°C to 45°C |
| Boiling Point | 171°C to 172°C |
| Flash Point | 77°C (170°F) |
| Quantity | 25 g |
| Beilstein | 1340501 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,7439 |
| Solubility Information | Soluble in hot water,alcohol,and diethyl ether. |
| Formula Weight | 118.18 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | H227-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| EINECSNumber | 200-933-5 |
| RTECSNumber | EK1720000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.