Learn More
Pinacolone, 97%, Thermo Scientific Chemicals
Description
Pinacolone is a precursor to triazolylpinacolone, triadimefon, paclobutrazol, uniconazole and metribuzin. Furthermore, it acts as an intermediate for biologically active products such as antibacterial, antifungal, antiviral and antituberculous products. It is an important ketone in organic chemistry and participates in condensation, hydrogenation and reductive amination reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pinacolone is a precursor to triazolylpinacolone, triadimefon, paclobutrazol, uniconazole and metribuzin. Furthermore, it acts as an intermediate for biologically active products such as antibacterial, antifungal, antiviral and antituberculous products. It is an important ketone in organic chemistry and participates in condensation, hydrogenation and reductive amination reactions.
Solubility
Miscible with water, chloroform, ethanol, ether, acetone and carbon tetrachloride.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | -53°C |
Density | 0.803 |
Boiling Point | 105°C to 106°C |
Flash Point | 5°C (41°F) |
Assay Percent Range | 97% |
Refractive Index | 1.397 |
Linear Formula | (CH3)3CC(O)CH3 |
Quantity | 500 g |
UN Number | UN1224 |
Merck Index | 14,7440 |
Show More |
Safety and Handling
Hazard Category | H225-H302 |
Hazard Statement | GHS H Statement H225-H302 Highly flammable liquid and vapor. Harmful if swallowed. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S. |
EINECSNumber | 200-920-4 |
RTECSNumber | EL7700000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.