Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pinacolone, 97%, Thermo Scientific Chemicals

Catalog No. p-7080664
Click to view available options
Quantity:
100 g
500 g

Pinacolone is a precursor to triazolylpinacolone, triadimefon, paclobutrazol, uniconazole and metribuzin. Furthermore, it acts as an intermediate for biologically active products such as antibacterial, antifungal, antiviral and antituberculous products. It is an important ketone in organic chemistry and participates in condensation, hydrogenation and reductive amination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pinacolone is a precursor to triazolylpinacolone, triadimefon, paclobutrazol, uniconazole and metribuzin. Furthermore, it acts as an intermediate for biologically active products such as antibacterial, antifungal, antiviral and antituberculous products. It is an important ketone in organic chemistry and participates in condensation, hydrogenation and reductive amination reactions.

Solubility
Miscible with water, chloroform, ethanol, ether, acetone and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-97-8
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008846
InChI Key PJGSXYOJTGTZAV-UHFFFAOYSA-N
Synonym pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone
PubChem CID 6416
IUPAC Name 3,3-dimethylbutan-2-one
SMILES CC(=O)C(C)(C)C

Specifications

Melting Point °C to 53°C
Density 0.803 g/mL
Boiling Point 105°C to 106°C
Flash Point 5°C (41°F)
Assay Percent Range 97%
Refractive Index 1.397
Linear Formula (CH3)3CC(O)CH3
Quantity 100 g
UN Number UN1224
Merck Index 14,7440
Solubility Information Miscible with water,chloroform,ethanol,ether,acetone and carbon tetrachloride.
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material Pinacolone
Show More Show Less
Hazard Category H225-H302
Hazard Statement GHS H Statement
H225-H302
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 200-920-4
RTECSNumber EL7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.