Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pinin Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10055315H
Description
Pinin Polyclonal antibody specifically detects Pinin in Human, Mouse samples. It is validated for Western BlotSpecifications
Pinin | |
Polyclonal | |
Western Blot | |
140 kDa nuclear and cell adhesion-related phosphoprotein, Desmosome-associated protein, Domain-rich serine protein, DRS protein, DRSP, DRSSR-like protein, Melanoma metastasis clone A protein, MEMA, neutrophil protein, Nuclear protein SDK3, pinin, pinin, desmosome associated protein, SDK3 | |
The immunogen recognized by this antibody maps to a region between residue 667 and the C-terminus (residue 717) of human Pinin, desmosome associated protein using the numbering given in entry NP_002678.2 (GeneID 5411). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
5411 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction