Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antistat Pink Antistatic Bag, Open Top

Pink Antistatic Bags dissipate a static charge to ground preventing static charge building up on the package or device. Used for non-ESD sensitive items destined for use in an EPA

$4.35 - $81.70

Specifications

Autoclavable Not Autoclavable
Closure Type None
Color Pink
For Use With (Application) Packaging of Non-ESD Sensitive Items used within an EPA
Includes Bag Only
View More Specs

Products 18
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
17-200-233
View Documents
Antistat
0010001
5 x 3 in.
Pack of 100 for $5.83
Only null left
Add to Cart
 
17-150-199
View Documents
Antistat
0010003
6 x 4 in.
Pack of 100 for $4.35
Only null left
Add to Cart
 
17-151-174
View Documents
Antistat
0010006
8 x 5 in.
Pack of 100 for $5.83
Only null left
Add to Cart
 
17-150-201
View Documents
Antistat
0010008
8 x 6 in.
Pack of 100 for $6.31
Only null left
Add to Cart
 
17-150-202
View Documents
Antistat
0010009
10 x 6 in.
Pack of 100 for $8.11
Only null left
Add to Cart
 
17-150-203
View Documents
Antistat
0010014
10 x 8 in.
Pack of 100 for $10.87
Only null left
Add to Cart
 
17-150-204
View Documents
Antistat
0010017
12 x 8 in.
Pack of 100 for $12.72
Only null left
Add to Cart
 
17-150-205
View Documents
Antistat
0010020
12 x 10 in.
Pack of 100 for $15.90
Only null left
Add to Cart
 
17-150-206
View Documents
Antistat
0010021
14 x 10 in.
Pack of 100 for $18.02
Only null left
Add to Cart
 
17-150-207
View Documents
Antistat
0010023
14 x 12 in.
Pack of 100 for $21.75
Only null left
Add to Cart
 
17-150-208
View Documents
Antistat
0010026
20 x 16 in.
Pack of 100 for $40.55
Only null left
Add to Cart
 
17-150-209
View Documents
Antistat
0010028
18 x 12 in.
Pack of 100 for $27.05
Only null left
Add to Cart
 
17-150-210
View Documents
Antistat
0010029
16 x 12 in.
Pack of 100 for $26.50
Only null left
Add to Cart
 
17-150-211
View Documents
Antistat
0010030
24 x 18 in.
Pack of 100 for $51.70
Only null left
Add to Cart
 
17-150-212
View Documents
Antistat
0010031
18 x 16 in.
Pack of 100 for $36.60
Only null left
Add to Cart
 
17-150-213
View Documents
Antistat
0010035
36 x 24 in.
Pack of 100 for $81.70
Only null left
Add to Cart
 
17-150-214
View Documents
Antistat
0010037
18 x 14 in.
Pack of 100 for $31.55
Only null left
Add to Cart
 
17-150-215
View Documents
Antistat
0010076
24 x 20 in.
Pack of 100 for $57.60
Only null left
Add to Cart
 
Description

Description

  • The Pink Antistatic Bags are formed using blow-molded polyethylene with antistatic additives
  • Soft texture and flexible
  • Surface resistance of 107 to <1011
  • Pink antistatic bags have the ability to dissipate a static charge to ground
  • Prevents static charge from building up on the package or device
  • The material is also antistatic and will not charge up when rubbed against other materials
  • Bag material allows visual identification of the items being stored
  • 300 gauge thickness
Specifications

Specifications

Not Autoclavable
Pink
Bag Only
75 μ
None
Packaging of Non-ESD Sensitive Items used within an EPA
LDPE
3 mil
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.