Learn More
AquaPhoenix Scientific Pink Feldspar
SCI_ED
Catalog No. S02891 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Product Type:
Mineral
Catalog No. Product Type
S02891 Mineral
1 options
Catalog No. S02891 Supplier AquaPhoenix Scientific Supplier No. EM5111B
Add to Cart
Add to Cart
Naturally pink feldspar with visible crystal structure—used to teach mineral properties, rock classification, and Earth science fundamentals.
  • Size range: 1 to 1.25 inches (approximate)
  • Exhibits characteristic pink hues and crystalline texture
  • Great for mineral identification, hands-on geology lessons, and earth science classrooms
  • Common rock-forming mineral found in igneous and metamorphic rocks
Specifications
Product Type Mineral