Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PINX1 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232265B
Description
PINX1 Polyclonal antibody specifically detects PINX1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
PINX1 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
FLJ20565, hepatocellular carcinoma-related putative tumor suppressor, Liver-related putative tumor suppressor, LPTLPinX1, LPTS67-11-3 protein, MGC8850, PIN2 interacting protein 1, PIN2/TERF1 interacting, telomerase inhibitor 1, PIN2/TERF1-interacting telomerase inhibitor 1, PIN2-interacting protein 1, Pin2-interacting protein X1, Protein 67-11-3, TRF1-interacting protein 1 | |
The immunogen this antibody was made to, maps to a region between residue 225 to 275 of human PIN2 Interacting Protein 1 using the numbering given in entry NP_060354.4 (GeneID 54984). | |
0.1 mL | |
Cancer | |
54984 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction