Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PINX1 Polyclonal Antibody
PINX1 Polyclonal Antibody
Supplier: Thermo Scientific PA598747
Description
PINX1 Polyclonal Antibody for Western Blot, ELISA

Specifications
PINX1 | |
Polyclonal | |
Unconjugated | |
PINX1 | |
2210403I16Rik; 2610028A01Rik; 67-11-3; 67-11-3 protein; AU024023; EMBL:ABO28828.1}; FLJ20565; hepatocellular carcinoma-related putative tumor suppressor; liver-related putative tumor suppressor; LPTL; Lpts; LPTS1; MGC8850; PIN2 interacting protein 1; PIN2/TERF1 interacting, telomerase inhibitor 1; PIN2/TERF1-interacting telomerase inhibitor 1; PIN2/TRF1-interacting protein; pin2-interact; PIN2-interacting protein 1; PIN2-interacting protein X1; Pinx1; pinx1 {ECO:0000312; Protein 67-11-3; RGD1566025; TRF1-interacting protein 1 | |
Rabbit | |
Protein G | |
RUO | |
54984, 72400 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96BK5, Q9CZX5 | |
PINX1 | |
Recombinant Human PIN2/TERF1-interacting telomerase inhibitor 1 protein (151-299AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction