Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PINX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103347
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103347 Supplier Invitrogen™ Supplier No. PA5103347
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PINX1.

Microtubule-binding protein essential for faithful chromosome segregation. Mediates TRF1 and TERT accumulation in nucleolus and enhances TRF1 binding to telomeres. Inhibits telomerase activity. May inhibit cell proliferation and act as tumor suppressor. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PINX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PINX1
Gene Accession No. Q96BK5, Q9CZX5
Gene Alias 2210403I16Rik; 2610028A01Rik; 67-11-3; 67-11-3 protein; AU024023; EMBL:ABO28828.1}; FLJ20565; hepatocellular carcinoma-related putative tumor suppressor; liver-related putative tumor suppressor; LPTL; Lpts; LPTS1; MGC8850; PIN2 interacting protein 1; PIN2/TERF1 interacting, telomerase inhibitor 1; PIN2/TERF1-interacting telomerase inhibitor 1; PIN2/TRF1-interacting protein; pin2-interact; PIN2-interacting protein 1; PIN2-interacting protein X1; Pinx1; pinx1 {ECO:0000312; Protein 67-11-3; RGD1566025; TRF1-interacting protein 1
Gene Symbols PINX1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PINX1(Accession Q96BK5), corresponding to amino acid residues T205-A255.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54984, 72400
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.