Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PINX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501727931 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7931 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7931 Supplier Proteintech Group Inc Supplier No. 123681AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Valosin containing protein (VCP), also designated TERA (for transitional endoplasmic reticulum ATPase), is a member of the AAA family of ATPases, which are involved in a variety of cellular activities. VCP is involved in a variety of membrane functions and in the regulation of the cell cycle. VCP associates with ubiquitinated IkB-a as well as with the 26S proteosome, indicating a potential role for VCP in the proteosome-mediated degradation of IkB-a. NVL (nuclear valosin-containing protein-like), also known as NVLp, is an 856 amino acid nuclear protein belonging to the AAA ATPase family. Implicated in ATP-dependent nuclear processes and ribosome synthesis, NVL exists as three alternatively spliced isoforms designated NVL isoform 1 (NVLp.2), NVL isoform 2 (NVLp.1) and NVL isoform 3. Widely expressed, NVL is found at highest levels in pancreas, retina, heart, skeletal muscle and placenta.

Specifications

Antigen PINX1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PINX1
Gene Accession No. A4L691, Q96BK5, Q9CZX5
Gene Alias LPTL, LPTS, PIN2 interacting protein 1, Pin2 interacting protein X1, PINX1, Protein 67 11 3, TRF1 interacting protein 1
Gene Symbols PINX1
Host Species Rabbit
Immunogen PINX1 Fusion Protein Ag3043
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305963, 54984, 72400
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.