Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PION Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521092
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521092 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521092 Supplier Invitrogen™ Supplier No. PA521092
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is EL4 cell lysate. PA5-21092 can be used with blocking peptide PEP-1206.

Accumulation of the amyloid-beta peptide (Abeta) in the cerebral cortex is a critical event in the pathogenesis of Alzheimer's disease. The beta-amyloid protein precursor (APP) is cleaved by one of two beta-secretases (BACE and BACE2), producing a soluble derivative of the protein and a membrane anchored 99 -amino acid carboxy-terminal fragment (C99). The C99 fragment serves as substrate for -gamma-secretase to generate the 4 kDa amyloid-beta peptide (Abeta), which is deposited in the Alzheimer's disease patients' brains. PION, or GSAP, selectively increases amyloid-beta production through a mechanism involving its interaction with both -gamma-secretase and the APP C-terminal fragment, suggesting that PION may be a potential therapeutic target for the treatment of Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PION
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene GSAP
Gene Accession No. A4D1B5, Q3TCV3
Gene Alias A530088I07Rik; gamma-secretase activating protein; gamma-secretase-activating protein; Gamma-secretase-activating protein 16 kDa C-terminal form; GSAP; GSAP-16K; pigeon homolog; Pion; Protein pigeon homolog; tcag7.1314
Gene Symbols GSAP
Host Species Rabbit
Immunogen A 19 amino acid peptide near the carboxy terminus of human PION.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212167, 311984, 54103
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.