Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ActivGrip™ Nitrile Coated Gloves with MicroFinish™ Grip

Catalog No. 19149615 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-149-615 Medium
19-149-614 Small
19-149-617 X-Large
19-149-616 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-149-615 Supplier PIP™ Supplier No. 56AG566/M
Add to Cart
Add to Cart
This item is not returnable. View return policy

MicroFinish™ surface treatment enhances grip in dry, wet, and oily conditions. Seamless cotton liner increases comfort and improves fit.

  • Revolutionary surface treatment enhances grip in dry, wet, and oily conditions
  • MicroFinish™ adjusts to different conditions by reacting like tiny suction cups to prevent liquid permeation
  • Seamless cotton liner increases comfort and improves fit
  • Strong protection against punctures and abrasions

Specifications

Coating Material Nitrile
Color Green
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Nitrile
Product Type Dip-coated Gloves
Size Medium
For Use With (Application) Fishing, Machine Operation, Metal Handling, and Oily Parts Assembly
Glove Cuff Style Open
Liner Material Cotton/Polyester Blend
Series 56-AG566
Yarn Gauge 13
Length (English) 11.8 in.
Length (Metric) 30 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.