Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ActivGrip™ Nitrile Coated Gloves with MicroFinish™ Grip and Attached PVC Sleeves

Catalog No. 19121221A Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-121-221A Small
19-121-221B Medium
19-121-221C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-121-221A Supplier PIP™ Supplier No. 56AG567/S

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Revolutionary surface treatment provides a superior grip in dry, wet, and oily conditions. Attached 25 in. PVC sleeves provide incidental splash protection all the way up the arms.

  • Revolutionary surface treatment for enhanced grip in dry, wet, and oily conditions
  • MicroFinish™ adjusts to different conditions by reacting like tiny suction cups to provide liquid permeation resistance
  • Seamless cotton liner improves fit for increased comfort
  • Extended clear PVC arm for incidental splash protection
  • Strong protection to punctures and abrasions

Specifications

Coating Material Nitrile
Color Green, Clear
Exterior Finish No Texture
Hand Specific
Material PVC
Product Type Dip-coated Gloves
Size Small
For Use With (Application) Oily Parts Assembly, Metal Handling, Fishing, and Machine Operation
Liner Material Cotton
Series 56-AG567
Length (English) Cuff 25 in.
Length (Metric) Cuff 63.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.