Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Ambi-dex™ Industrial Grade Nitrile Gloves

Catalog No. 19121196C Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-121-196C Large
19-121-196B Medium
19-121-196A Small
19-121-196D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-121-196C Supplier PIP™ Supplier No. 63332/L/DEL
This item is not returnable. View return policy

For use in a broad range of applications for light hand protection from liquids, chemicals and debris

  • Economical puncture resistance with great comfort, dexterity and tactile sensitivity
  • Contains no natural rubber latex proteins
  • Ambidextrous
  • Made from component materials that comply with Federal Regulations for food contact 21CFR, 170-199
  • Blue Color (except 63-732PF which is black)

Specifications

Certifications/Compliance FDA 21 CFR 170-199, ISO 9002
Color Blue
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Large
Thickness (English) 4 mil
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.