Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ArmorLite™ Textured Nitrile Coated Cotton Gloves

Catalog No. 19142756 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-142-756 Medium
19-142-757 Large
19-142-758 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-142-756 Supplier PIP™ Supplier No. 563170/M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves feature a thin nitrile coating with a textured finish for enhanced grip capabilities, while ActiFresh™ treatment reduces germs and odors.

  • Cotton interlock liner thinly coated with blue nitrile
  • Comfortable and flexible for a high degree of dexterity
  • Textured coating provides enhanced grip capabilites
  • Effective, affordable protection from physical hazards such as cut, puncture, snags and abrasions
  • Resistant to grease and oils
  • Made in ISO 9001:2000 / ISO 14001:2000 facilities
  • Launderable

Specifications

Coated Area Palm, Fingers, Knuckles
Coating Material Nitrile
Color Blue, Natural
Exterior Finish Partially Textured
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size Medium
For Use With (Application) General Handling, Assembly, Fabrication, Inspection, and Maintenance
Glove Cuff Style Knit
Series 56-3170
Length (English) 9.8 in.
Length (Metric) 25 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.