Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ 52-3672 Unsupported Neoprene Coated Latex Gloves with Raised Diamond Grip

Catalog No. 19161646B Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-161-646B Medium
19-161-646A Small
19-161-646C Large
19-161-646D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-161-646B Supplier PIP™ Supplier No. 523672/M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Double-dipped neoprene construction provides protection against a wide range of chemicals such as acids, caustics, solvents, and oils. Raised diamond pattern enhances grip capabilities in wet or oily conditions.

  • Neoprene coating provides protection from a wide range of chemicals such as acids, caustics, solvents, and oils
  • Double dipped construction combines the versatile chemical resistance properties of neoprene with the comfort of latex
  • Straight cuff adds additional length to protect the forearm from liquid run-off
  • Flock lining increases comfort and eases donning/doffing
  • Raised diamond pattern enhances grip capabilities
  • All component materials comply with Federal regulations for food contact 21 CFR, 170-199

Specifications

Certifications/Compliance 21 CFR, 170-199
Coating Material Neoprene
Color Blue, Yellow
Exterior Finish Partially Textured
Hand Specific
Latex Free Yes
Material Latex
Product Type Dip-coated Gloves
Size Medium
Thickness (English) 19 mil
EN388 Standard 1X11
For Use With (Application) General Maintenance, Janitorial Work, Petrochemical, and Photo Finishing
Glove Cuff Style Straight
Liner Material Flocked Cotton
Series 52-3672
Length (English) 12.6 in.
Length (Metric) 32 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.