Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ GX3™ Dyneema™ Diamond Blended Gloves, Polyurethane Coating

Catalog No. 19824129 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Blue
Brown
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-824-129 2X-Large Blue
19-824-112 Small Red
19-824-113 Medium Yellow
19-824-114 Large Brown
19-824-115 X-Large Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-824-129 Supplier PIP™ Supplier No. 19D320/XXL
This item is not returnable. View return policy

Lightweight shell with polyurethane coating provides dexterity, tactile sensitivity and ANSI A3 cut resistance. Gloves are also washable, resistant to chemicals, water and ultraviolet light.

  • Seamless knit, Dyneema™ diamond blended
  • Polyurethane coated smooth grip
  • Washable, resistant to chemicals, water and UV light
  • Color coded cuff hems for easy identification
  • Puncture and abrasion resistance
  • Cut-resistant and tactile sensitivity
  • Applications include electronics, glass, metal, food processing

Certifications

The Oeko-Tex™ Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin.

Specifications

Certifications/Compliance (Europe) CE
Construction Seamless Knit
Coating Material Polyurethane
Color Salt and Pepper, Gray
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free Yes
Material Dyneema
Product Type Cut-resistant Gloves
Size 2X-Large
EN388 Standard EN 388:4442
For Use With (Application) Electronics, glass cutting, metal handling, food and meat processing
Glove Cuff Color Blue
Glove Cuff Style Knit
Model 19-D320/XXL
Series G-Tek™ 3GX™
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.